Learn More
(1-Methyl-2-phenylethyl)-carbamic Acid, 2-Methylpropyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (1-Methyl-2-phenylethyl)-carbamic Acid, 2-Methylpropyl Ester |
| CAS | 27822-62-4 |
| Molecular Formula | C14H21NO2 |
| Quantity | 5 mg |
| Synonym | Isobutyl(1-phenylpropan-2-yl)carbamate |
| InChI Formula | InChI=1S/C14H21NO2/c1-11(2)10-17-14(16)15-12(3)9-13-7-5-4-6-8-13/h4-8,11-12H,9-10H2,1-3H3,(H,15,16) |
| SMILES | CC(C)COC(=O)NC(C)Cc1ccccc1 |
| IUPAC Name | 2-methylpropyl N-(1-phenylpropan-2-yl)carbamate |
| Molecular Weight (g/mol) | 235.32 |
| Formula Weight | 235.1572 |