Learn More
(1-Methyl-2-phenylethyl)-carbamic Acid, 2-Methylpropyl Ester, TRC
Product Code. 30465892
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465892 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30465892 Supplier Toronto Research Chemicals Supplier No. TRCM33892350MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1-Methyl-2-phenylethyl)-carbamic Acid, 2-Methylpropyl Ester
CAS 27822-62-4
Molecular Formula C14H21NO2
Quantity 50 mg
Synonym Isobutyl(1-phenylpropan-2-yl)carbamate
InChI Formula InChI=1S/C14H21NO2/c1-11(2)10-17-14(16)15-12(3)9-13-7-5-4-6-8-13/h4-8,11-12H,9-10H2,1-3H3,(H,15,16)
SMILES CC(C)COC(=O)NC(C)Cc1ccccc1
IUPAC Name 2-methylpropyl N-(1-phenylpropan-2-yl)carbamate
Molecular Weight (g/mol) 235.32
Formula Weight 235.1572