Learn More
1-Methyl-2-(methylthio)imidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-2-(methylsulfanyl)-1H-imidazole |
| CAS | 14486-52-3 |
| Molecular Formula | C5 H8 N2 S |
| Quantity | 250 mg |
| Synonym | 1-Methyl-2-(methylsulfanyl)-1H-imidazole, 1-Methyl-2-(methylsulphanyl)-1H-imidazole, 1H-Imidazole, 1-methyl-2-(methylthio)- (9CI), Imidazole, 1-methyl-2-(methylthio)- (8CI), 1-Methyl-2-(methylthio)imidazole, 2-Methylthio-3-methylimidazole |
| InChI Formula | InChI=1S/C5H8N2S/c1-7-4-3-6-5(7)8-2/h3-4H,1-2H3 |
| SMILES | CSc1nccn1C |
| IUPAC Name | 1-methyl-2-methylsulfanylimidazole |
| Molecular Weight (g/mol) | 128.2 |
| Formula Weight | 128.0408 |