missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester, 95%, Thermo Scientific™

Product Code. p-7030157
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15467715

Brand: Thermo Scientific Alfa Aesar H27703.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Important intermediates and reagents in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1044851-76-4
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.041
MDL Number MFCD07777079
InChI Key GJNNJJRONITJEY-UHFFFAOYSA-N
Synonym 5-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazole-5-boronic acid neopentyl glycol ester, 1-methyl-1h-pyrazole-5-boronic acid, neopentyl glycol ester, 5-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-1-methylpyrazole, acmc-20am84, 5-5,5-dimethyl-1,3,2 dioxaborinan-2-yl-1-methyl-1h-pyrazole, 5-5,5-dimethyl-1,3,2-dioxaborinane-2-yl-1-methyl-1h-pyrazole
PubChem CID 44119444
IUPAC Name 5-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-1-methylpyrazole
SMILES B1(OCC(CO1)(C)C)C2=CC=NN2C

Specifications

Melting Point 72°C to 75°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 194.04
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.