Learn More
Wheaton™ 1 μL Inoculation Loop
Click to view available options
Size:
Short
Standard
Unit Size:
Pack of 6000
Description
- Meets quantitative standards for management of microbiological samples
- For accurate liquid transfer and plating
- Calibrated
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Length (Metric) | 170 mm |
| Material | Polystyrene |
| Sterility | Sterile |
| Size | Short |
| Color | Clear |
| Quantity | 6000 |
| Packaging Type | Packaged in peel pouch |
| Calibrated | Yes |
| Show More |