Learn More
Hamilton™ 1 μL 7000 Series Microliter Syringe
Product Code. 17804212
Click to view available options
Needle Gauge:
22 G
23s G
25 G
26s G
Needle Point Style:
Blunt
Cone
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Knurled Hub
Knurled Hub, Removable
Unit Size:
Each
Product Code. 17804212
Brand: Hamilton™ CAL80176
Please to purchase this item. Need a web account? Register with us today!
The 7000 series syringes are modified Microliter syringes with a plunger-in-needle design.
  • The only zero-dead volume syringe available.
  • The fine gauge tungsten plunger wire extends to the tip of the needle, ensuring that all solvent is expelled, dispensing volumes up to 1 μL.
Specifications
Autoclavable Autoclavable
Material Tungsten (Barrel)
Needle Gauge 23s G
Diameter (English) Inner 0.0057 in.
Diameter (English) Inner Needle 0.0057 in.
Diameter (Metric) Inner Needle 0.145 mm
Diameter (English) Outer Needle 0.020 in.
Diameter (English) Outer 0.308 in.
Diameter (Metric) Inner 0.145 mm
Diameter (Metric) Outer 7.82 mm
Diameter (Metric) Outer Needle 0.515 mm
Length (English) Needle 1.71 in.
Product Type Microliter Syringe
Max. Temperature (English) 239°F
Max. Temperature (Metric) 115°C
Min. Temperature (English) 50°F
Model 7001 KH
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 1 μL
Length (Metric) Needle 43.4 mm
Needle Point Style Cone
Connection Type Cemented