Learn More
STEMCELL Technologies 1 kb DNA Ladder
Product Code. 17169901
Encompass_Preferred
Change view
Click to view available options
Quantity:
600 μL
Unit Size:
600µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17169901 600 μL 600µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17169901 Supplier STEMCELL Technologies Supplier No. 79017
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
DNA ladder consisting of 13 fragments from 250 - 10,000 base pairs for determining the size of double-stranded DNA by gel electrophoresis
  • 1 kb DNA Ladder allows for determining the size of double-stranded DNA from 250 - 10000 base pairs (bp) by gel electrophoresis
  • The ladder consists of 13 double-stranded, blunt-end DNA fragments, of sizes 250, 500, 750, 1000, 1500, 2000, 2500, 3000, 4000, 5000, 6000, 8000, and 10,000 bp (see Figure 1)
  • The 1000 and 3000 bp fragments have greater intensity relative to the other bands for easy identification
  • DNA Ladder is not intended for quantitative analysis
  • The 1 kb DNA ladder comes pre-mixed with DNA Loading Dye, which is also available for individual sale (Catalog #79018), for loading DNA samples into agarose gel wells
  • The recommended loading volume of the 1 kb DNA ladder is 6 μL per lane
Specifications
For Use With (Application) Disease Modeling
Quantity 600 μL