Learn More
1-Iodooctane-1,1-d2, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Iodooctane-1,1-D2 |
| CAS | 89232-08-6 |
| Molecular Formula | C8 D2 H15 I |
| Quantity | 0.25 g |
| Synonym | Octane-1,1-d2, 1-iodo- (9 CI), 1-Iodooctane-1,1-d2, 1-Iodooctane-1,1-D2 |
| InChI Formula | InChI=1 S/C8H17I/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3/i8D2 |
| SMILES | [2 H]C([2 H])(I)CCCCCCC |
| IUPAC Name | 1,1-dideuterio-1-iodooctane |
| Molecular Weight (g/mol) | 242.1374 |
| Formula Weight | 242.05 g/mol |
| Show More |