Learn More
1-Hydroxyadamantane-d15, CDN
Product Code. 30317348
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317348 0.01 g 0.01g
30319357 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317348 Supplier CDN Supplier No. CDND79430.01G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Adamantanol-D15
CAS 33830-11-4
Molecular Formula C10 D15 H O
Quantity 0.01 g
Synonym 1-Adamantan-d15-ol (8 CI), 1-Hydroxyadamantane-d15, 1-Adamantanol-D15
InChI Formula InChI=1 S/C10H16O/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9,11 H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7 D,8 D,9 D
SMILES [2 H]C1([2 H])C2([2 H])C([2 H])([2 H])C3([2 H])C([2 H])([2 H])C1([2 H])C([2 H])([2 H])C(O)(C3([2 H])[2 H])C2([2 H])[2 H]
IUPAC Name 2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantan-1-ol
Molecular Weight (g/mol) 167.3259
Formula Weight 167.2143 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity