Learn More
1-(Heptafluorobutyryl)imidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(Heptafluorobutyryl)imidazole |
| CAS | 32477-35-3 |
| Molecular Formula | C7 H3 F7 N2 O |
| Quantity | 5 g |
| Synonym | 2,2,3,3,4,4,4-Heptafluoro-1-(1H-imidazol-1-yl)-1-butanone, 1-(2,2,3,3,4,4,4-Heptafluoro-1-oxobutyl)-1H-imidazole, NSC 151966 |
| InChI Formula | InChI=1S/C7H3F7N2O/c8-5(9,6(10,11)7(12,13)14)4(17)16-2-1-15-3-16/h1-3H |
| SMILES | FC(F)(F)C(F)(F)C(F)(F)C(=O)n1ccnc1 |
| IUPAC Name | 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one |
| Molecular Weight (g/mol) | 264.1 |
| Formula Weight | 264.0134 |