Learn More
1-(Heptafluorobutyryl)imidazole, 97%
Product Code. 15219195
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15219195 25 g 25g
11392705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15219195 Supplier Thermo Scientific Alfa Aesar Supplier No. L00221.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 32477-35-3 | C7H3F7N2O | 264.103 g/mol
1-(Heptafluorobutyryl)imidazole is used in gas chromatography for the determination of various pharmaceutical compounds like retronecine in biological matrixes. It acts as a reagent for derivatize amine-groups. It plays an essential role to reduce GC column degradation due to its non-acidic property. It is an acylating agent, which converts thermally unstable compounds to its more volatile derivative and then made suitable for GC analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(Heptafluorobutyryl)imidazole is used in gas chromatography for the determination of various pharmaceutical compounds like retronecine in biological matrixes. It acts as a reagent for derivatize amine-groups. It plays an essential role to reduce GC column degradation due to its non-acidic property. It is an acylating agent, which converts thermally unstable compounds to its more volatile derivative and then made suitable for GC analysis.
Solubility
Miscible with chloroform and methanol.
Notes
Hygroscopic and air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32477-35-3
Molecular Formula C7H3F7N2O
Molecular Weight (g/mol) 264.103
MDL Number MFCD00014503
InChI Key MSYHGYDAVLDKCE-UHFFFAOYSA-N
Synonym n-heptafluorobutyrylimidazole, 1-heptafluorobutyryl imidazole, heptafluorobutyrylimidazole, n-heptafluorobutyryl imidazole, hfbi, 1-perfluorobutyryl imidazole, 2,2,3,3,4,4,4-heptafluoro-1-1h-imidazol-1-yl butan-1-one, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl-1h-imidazole, 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-yl butan-1-one, 1h-imidazole, 1-2,2,3,3,4,4,4-heptafluoro-1-oxobutyl
PubChem CID 94431
IUPAC Name 2,2,3,3,4,4,4-heptafluoro-1-imidazol-1-ylbutan-1-one
SMILES C1=CN(C=N1)C(=O)C(C(C(F)(F)F)(F)F)(F)F
Specifications
Melting Point 10°C to 13°C
Density 1.49
Boiling Point 160°C to 162°C
Flash Point 77°C (170°F)
Refractive Index 1.385
Quantity 25 g
Beilstein 4488026
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform and methanol.
Formula Weight 264.1
Percent Purity ≥98%
Chemical Name or Material 1-(Heptafluorobutyryl)imidazole