Learn More
1-Ethyl-4-iodobenzene, 98%
Product Code. 11448106
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448106 10 g 10g
11458106 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11448106 Supplier Thermo Scientific Alfa Aesar Supplier No. A14138.09
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
CAS: 25309-64-2 | C8H9I | 232.064 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 25309-64-2
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00060663
InChI Key OOLSRHZMXAYDFB-UHFFFAOYSA-N
Synonym 4-ethyliodobenzene, benzene, 1-ethyl-4-iodo, pubchem3877, 1-iodo-4-ethylbenzene, acmc-1chvo, p-ethyliodobenzene, 1-ethyl-4-iodo-benzene, #, 1-ethyl-4-iodanyl-benzene
PubChem CID 141189
IUPAC Name 1-ethyl-4-iodobenzene
SMILES CCC1=CC=C(C=C1)I
Spezifikation
Melting Point -17°C
Density 1.6
Boiling Point 112°C to 113°C (20 mmHg)
Flash Point 107°C (224°F)
Odor Mild
Refractive Index 1.591
Quantity 10 g
Beilstein 2076973
Sensitivity Light sensitive
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 1-Ethyl-4-iodobenzene