missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-fluorobenzene, 97%, Thermo Scientific™

Product Code. 11382296
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11382296

Brand: Thermo Scientific Alfa Aesar L05940.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Ethyl-4-fluorobenzene is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-47-2
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.16
MDL Number MFCD00039218
InChI Key BLDNWXVISIXWKZ-UHFFFAOYSA-N
Synonym p-ethylfluorobenzene, 4-ethylfluorobenzene, ethyl-4-fluorobenzene, p-fluoroethylbenzene, benzene, 1-ethyl-4-fluoro, 4-fluoroethylbenzene, p-ethyl fluorobenzene, 4-fluoro-ethylbenzene, 1,4-ethylfluorobenzene, pubchem22752
PubChem CID 136303
IUPAC Name 1-ethyl-4-fluorobenzene
SMILES CCC1=CC=C(F)C=C1

Specifications

Boiling Point 139°C
Flash Point 30°C (86°F)
Refractive Index 1.471
Quantity 25 g
UN Number UN1993
Beilstein 1856987
Solubility Information Not miscible in water.
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 1-Ethyl-4-fluorobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.