Learn More
1-Deoxy-1-(L-aspartyl)-D-fructose Ammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Deoxy-1-(L-aspartyl)-D-fructose Ammonium Salt |
| CAS | 31105-02-9 |
| Molecular Formula | C10 H17 N O9 |
| Quantity | 10 mg |
| Synonym | (S)-1-Deoxy-1-[(1,2-dicarboxyethyl)amino]-D-fructose, 1-deoxy-1-[(1,2-dicarboxyethyl)amino]-fructose, D-1-Deoxy-1-[(L-1,2-dicarboxyethyl)amino]-fructose, 1-Deoxy-1-(L-aspartic acid)-D-fructose, 1-Deoxy-1-(L-aspartyl)-D-fructose, 1-N-L-Aspartic Acid 1-deoxy-D-fructose, Fructose-aspartic acid, N-(1-Deoxy-D-fructosyl)-L-aspartic acid |
| InChI Formula | InChI=1S/C10H17NO9/c12-3-6(14)9(18)8(17)5(13)2-11-4(10(19)20)1-7(15)16/h4,6,8-9,11-12,14,17-18H,1-3H2,(H,15,16)(H,19,20)/t4-,6+,8+,9+/m0/s1 |
| SMILES | OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CN[C@@H](CC(=O)O)C(=O)O |
| IUPAC Name | (2S)-2-[[(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexyl]amino]butanedioic acid |
| Molecular Weight (g/mol) | 295.243 |
| Formula Weight | 295.09 |