missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)naphthalene, 94%, Thermo Scientific Chemicals

Product Code. 11464250
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
This item is not returnable. View return policy

Product Code. 11464250

Brand: Thermo Scientific Alfa Aesar B24385.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

1-(Chloromethyl)naphthalene is a key material for synthesis dye pigment and as fluorescent brightening agent. It is employed in synthetic resin and medicine. Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives produce ortho- or para-substituted carbocycles in satisfactory to excellent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Chloromethyl)naphthalene is a key material for synthesis dye pigment and as fluorescent brightening agent. It is employed in synthetic resin and medicine. Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives produce ortho- or para-substituted carbocycles in satisfactory to excellent yields.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-52-2
Molecular Formula C11H9Cl
Molecular Weight (g/mol) 176.643
MDL Number MFCD00004042
InChI Key XMWGTKZEDLCVIG-UHFFFAOYSA-N
Synonym 1-chloromethyl naphthalene, naphthalene, 1-chloromethyl, 1-naphthylmethyl chloride, 1-menaphthyl chloride, 1-chlormethyl naftalen, chloromethyl naphthalene, 1-chloro methylnaphthalene, alpha-naphthylmethyl chloride
PubChem CID 6845
IUPAC Name 1-(chloromethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCl

Specifications

Melting Point 31°C to 33°C
Boiling Point 291°C to 292°C
Flash Point 132°C (270°F)
Odor Pungent
Quantity 500 g
UN Number UN3261
Beilstein 636885
Formula Weight 176.65
Percent Purity 94%
Chemical Name or Material 1-(Chloromethyl)naphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.