Learn More
1-Chloroisofluorane, TRC
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane |
| CAS | 32778-08-8 |
| Molecular Formula | C3 H Cl2 F5 O |
| Quantity | 1 g |
| Synonym | (2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane, Isoflurane Imp. C (EP) |
| InChI Formula | InChI=1S/C3HCl2F5O/c4-1(2(6,7)8)11-3(5,9)10/h1H |
| SMILES | FC(F)(F)C(Cl)OC(F)(F)Cl |
| IUPAC Name | 2-chloro-2-[chloro(difluoro)methoxy]-1,1,1-trifluoroethane |
| Molecular Weight (g/mol) | 218.94 |
| Formula Weight | 217.9325 |