Learn More
1-Chloro-4-(chlorophenylmethyl)benzene, TRC
Product Code. 30007487
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30007487 Supplier Toronto Research Chemicals Supplier No. TRCC3648555G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 134-83-8
Molecular Formula C13 H10 Cl2
Molecular Weight (g/mol) 237.12
Synonym Chloro(p-chlorophenyl)phenylmethane, 1-Chloro-4-(chloro(phenyl)methyl)benzene, 4-Chlorobenzhydryl Chloride, Chloro(p-chlorophenyl)phenylmethane, NSC 49126, p-Chlorobenzhydryl Chloride
IUPAC Name 1-chloro-4-[chloro(phenyl)methyl]benzene
SMILES ClC(c1ccccc1)c2ccc(Cl)cc2
Specifications
Quantity 5 g
InChI Formula InChI=1S/C13H10Cl2/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,13H
Formula Weight 236.016
Chemical Name or Material 1-Chloro-4-(chlorophenylmethyl)benzene