Learn More
((1-Chloro-2-methylpropan-2-yl)oxy)trimethylsilane, TRC
Specifications
Specifications
| Chemical Name or Material | ((1-Chloro-2-methylpropan-2-yl)oxy)trimethylsilane |
| CAS | 31557-15-0 |
| Molecular Formula | C7H17ClOSi |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C7H17ClOSi/c1-7(2,6-8)9-10(3,4)5/h6H2,1-5H3 |
| SMILES | CC(C)(CCl)O[Si](C)(C)C |
| IUPAC Name | (1-chloro-2-methylpropan-2-yl)oxy-trimethylsilane |
| Molecular Weight (g/mol) | 180.75 |