missing translation for 'onlineSavingsMsg'
Learn More

1-Butoxy-4-eth-1-ynylbenzene, 97%, Thermo Scientific™

Product Code. p-2695655
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10572262

Brand: Thermo Scientific Maybridge BTB09896DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 79887-15-3
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00173878
InChI Key JMLXBCSKFMMFGF-UHFFFAOYSA-N
Synonym 1-butoxy-4-eth-1-ynylbenzene, benzene,1-butoxy-4-ethynyl, 4-n-butoxyphenylacetylene, 4-butoxyphenylacetylene, 1-ethynyl-4-butoxybenzene, 1-butoxy-4-ethynyl-benzene
PubChem CID 2775120
IUPAC Name 1-butoxy-4-ethynylbenzene
SMILES CCCCOC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)3OC6H4Cidentical toCH
Quantity 1 g
Formula Weight 174.24
Percent Purity 97%
Chemical Name or Material 1-Butoxy-4-eth-1-ynylbenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.