Learn More
1-Butan-1,1-d2-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Butanol-1,1-D2 |
| CAS | 32586-14-4 |
| Molecular Formula | C4H8D2O |
| Quantity | 100 mg |
| Synonym | 1,1-Dideuteriobutan-1-ol |
| InChI Formula | InChI=1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i4D2 |
| SMILES | OC([2H])([2H])CCC |
| IUPAC Name | butan-1,1-d2-1-ol |
| Molecular Weight (g/mol) | 76.13 |
| Formula Weight | 76.09 |