Learn More
1-Bromopropane-3,3,3-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopropane-3,3,3-d3 |
| CAS | 61809-88-9 |
| Molecular Formula | C3 D3 H4 Br |
| Quantity | 0.5 g |
| Synonym | 1-Propyl Bromide-d3, 3-Bromopropane-d3, Abzol JG-d3, Ascusol MC-d3, Drysolv-d3, Leksol-d3, Propyl Bromide-d3, n-Propyl Bromide-d3; |
| InChI Formula | InChI=1 S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCBr |
| IUPAC Name | 3-bromo-1,1,1-trideuteriopropane |
| Molecular Weight (g/mol) | 126.01 |
| Formula Weight | 124.9919 g/mol |
| Show More |