Learn More
1-Bromoheptane-7,7,7-d3, CDN
Product Code. 30311562
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311562 0.5 g 0.5g
30311945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311562 Supplier CDN Supplier No. CDND52150.5G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromoheptane-7,7,7-D3
CAS 344253-18-5
Molecular Formula C7 D3 H12 Br
Quantity 0.5 g
Synonym Heptane-1,1,1-d3, 7-bromo- (9 CI), 7-Bromoheptane-1,1,1-d3, 1-Bromoheptane-7,7,7-d3, 1-Bromoheptane-7,7,7-D3
InChI Formula InChI=1 S/C7H15Br/c1-2-3-4-5-6-7-8/h2-7H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])CCCCCCBr
IUPAC Name 7-bromo-1,1,1-trideuterioheptane
Molecular Weight (g/mol) 182.1165
Formula Weight 181.0545 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity