Learn More
1-Bromodecane-10,10,10-d3, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Bromodecane-10,10,10-D3 |
| CAS | 284474-47-1 |
| Molecular Formula | C10 D3 H18 Br |
| Quantity | 0.25 g |
| Synonym | Decane-1,1,1-d3, 10-bromo- (9 CI), 1-Bromodecane-d3, 1-Bromodecane-10,10,10-D3, 10-Bromodecane-1,1,1-d3, 1-Bromodecane-10,10,10-d3, 1-Decyl-10,10,10-d3 bromide |
| InChI Formula | InChI=1 S/C10H21Br/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCBr |
| IUPAC Name | 10-bromo-1,1,1-trideuteriodecane |
| Molecular Weight (g/mol) | 224.1962 |
| Formula Weight | 223.1015 g/mol |
| Show More |