Learn More
1-Bromodecane-1,1,2,2-d4, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Bromodecane-1,1,2,2-D4 |
| CAS | 1219805-58-9 |
| Molecular Formula | C10 D4 H17 Br |
| Quantity | 0.5 g |
| Synonym | 1-Bromodecane-d4, 1-Bromodecane-1,1,2,2-D4, 1-Bromodecane-1,1,2,2-d4, 1-Decyl-1,1,2,2-d4 bromide |
| InChI Formula | InChI=1 S/C10H21Br/c1-2-3-4-5-6-7-8-9-10-11/h2-10H2,1H3/i9D2,10D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])CCCCCCCC |
| IUPAC Name | 1-bromo-1,1,2,2-tetradeuteriodecane |
| Molecular Weight (g/mol) | 225.2024 |
| Formula Weight | 224.1078 g/mol |
| Show More |