Learn More
1-Bromo-6-(trimethylammonium)hexyl Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-6-(trimethylammonium)hexyl Bromide |
| CAS | 32765-81-4 |
| Molecular Formula | C9 H21 Br N . Br |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H21BrN.BrH/c1-11(2,3)9-7-5-4-6-8-10;/h4-9H2,1-3H3;1H/q+1;/p-1 |
| SMILES | [Br-].C[N+](C)(C)CCCCCCBr |
| IUPAC Name | 6-bromohexyl(trimethyl)azanium;bromide |
| Molecular Weight (g/mol) | 303.08 |
| Formula Weight | 301.0041 |