missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-iodobenzene, 98+%, stab. with copper
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
2-Bromoiodobenzene is used in the synthesis of diarylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoiodobenzene is used in the synthesis of diarylamines.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
2-Bromoiodobenzene is used in the synthesis of diarylamines.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 9°C to 10°C |
| Density | 2.256 |
| Boiling Point | 257°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.6645 |
| Quantity | 25 g |
| Beilstein | 1855309 |
| Sensitivity | Light sensitive |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 282.91 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction