missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-cyanopiperidine, 96%, Thermo Scientific™

Product Code. p-7030049
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483717 250 mg 250mg
11436480 1 g 1g
11446480 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15483717 Supplier Thermo Scientific Alfa Aesar Supplier No. H27580.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91419-52-2
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.277
MDL Number MFCD01861223
InChI Key UQADQTBQNVARAP-UHFFFAOYSA-N
Synonym 1-boc-4-cyanopiperidine, 1-n-boc-4-cyanopiperidine, n-boc-4-cyanopiperidine, 1-n-boc-4-cyano-piperidine, n-boc-4-cyano-piperidine, 1-piperidinecarboxylic acid, 4-cyano-, 1,1-dimethylethyl ester, boc-4-cyanopiperidine, 4-cyano-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-cyano-1-piperidinecarboxylate, n-boc-piperidine-4-carbonitrile
PubChem CID 1514443
IUPAC Name tert-butyl 4-cyanopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C#N

Specifications

Melting Point 45°C to 48°C
Quantity 250 mg
UN Number UN3439
Formula Weight 210.27
Percent Purity 96%
Chemical Name or Material 1-Boc-4-cyanopiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.