Learn More
1-Benzylideneaminohydantoin, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Benzylideneaminohydantoin |
| CAS | 2827-57-8 |
| Molecular Formula | C10 H9 N3 O2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C10H9N3O2/c14-9-7-13(10(15)12-9)11-6-8-4-2-1-3-5-8/h1-6H,7H2,(H,12,14,15)/b11-6+ |
| SMILES | O=C1CN(\N=C\c2ccccc2)C(=O)N1 |
| IUPAC Name | 1-[(E)-benzylideneamino]imidazolidine-2,4-dione |
| Molecular Weight (g/mol) | 203.2 |
| Formula Weight | 203.0695 |