Learn More
1-benzyl-3,3-dimethylthiourea, TRC
Specifications
Specifications
| Chemical Name or Material | 1-benzyl-3,3-dimethylthiourea |
| CAS | 2741-14-2 |
| Molecular Formula | C10H14N2S |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C10H14N2S/c1-12(2)10(13)11-8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3,(H,11,13) |
| SMILES | CN(C)C(=S)NCC1=CC=CC=C1 |
| IUPAC Name | 3-benzyl-1,1-dimethylthiourea |
| Molecular Weight (g/mol) | 194.3 |
| Formula Weight | 194.0878 |