Learn More
1-benzyl-3,3-dimethylthiourea, TRC
Product Code. 30342681
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342681 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30342681 Supplier Toronto Research Chemicals Supplier No. TRCB53700025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-benzyl-3,3-dimethylthiourea
CAS 2741-14-2
Molecular Formula C10H14N2S
Quantity 25 mg
InChI Formula InChI=1S/C10H14N2S/c1-12(2)10(13)11-8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3,(H,11,13)
SMILES CN(C)C(=S)NCC1=CC=CC=C1
IUPAC Name 3-benzyl-1,1-dimethylthiourea
Molecular Weight (g/mol) 194.3
Formula Weight 194.0878