Learn More
1-Benzoyl-4-(2,3-dichlorophenyl)piperazine, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Benzoyl-4-(2,3-dichlorophenyl)piperazine |
| CAS | 902456-03-5 |
| Molecular Formula | C21 H26 O9 S |
| Quantity | 25 mg |
| Synonym | [4-(2,3-Dichlorophenyl)-1-piperazinyl]phenylmethanone |
| InChI Formula | InChI=1S/C21H26O9S/c1-11-6-8-16(9-7-11)31-21-20(29-15(5)25)19(28-14(4)24)18(27-13(3)23)17(30-21)10-26-12(2)22/h6-9,17-21H,10H2,1-5H3/t17?,18-,19?,20?,21-/m0/s1 |
| SMILES | CC(=O)OCC1O[C@@H](Sc2ccc(C)cc2)C(OC(=O)C)C(OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | [(3S,6S)-3,4,5-triacetyloxy-6-(4-methylphenyl)sulfanyloxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 454.491 |
| Formula Weight | 454.13 |