Learn More
1-Azido-4-(bromomethyl)benzene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Azido-4-(bromomethyl)benzene |
| CAS | 74489-49-9 |
| Molecular Formula | C7H6N3Br |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C7H6BrN3/c8-5-6-1-3-7(4-2-6)10-11-9/h1-4H,5H2 |
| SMILES | BrCc1ccc(cc1)N=[N+]=[N-] |
| IUPAC Name | 1-azido-4-(bromomethyl)benzene |
| Molecular Weight (g/mol) | 212.05 |
| Formula Weight | 210.9745 |