Learn More
1-Azakenpaullone, MedChemExpress
Product Code. 16488158
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
Product Code. 16488158
Brand: Medchem Express HY59090
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress 1-Azakenpaullone (1-Akp) is a highly selective and ATP-competitive inhibitor of glycogen synthase kinase-3 β (GSK-3β), with an IC50 value of 18 nM.
1-Azakenpaullone (1-Akp) is a highly selective and ATP-competitive inhibitor of glycogen synthase kinase-3 β (GSK-3β), with an IC50 value of 18 nM.
Specifications
Chemical Name or Material 1-Azakenpaullone
CAS 676596-65-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H10BrN3O
Quantity 10 mg
Synonym 1-Akp
Solubility Information DMSO : 16.67 mg/mL (50.80 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CN=C12)CC3=C2NC4=C3C=C(Br)C=C4
Molecular Weight (g/mol) 328.16
Formula Weight 328.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism