Learn More
1-Amino-2-naphthol Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Amino-2-naphthol Hydrochloride |
| CAS | 1198-27-2 |
| Molecular Formula | C10H9NO . HCl |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C10H9NO.ClH/c11-10-8-4-2-1-3-7(8)5-6-9(10)12;/h1-6,12H,11H2;1H |
| SMILES | Cl.Nc1c(O)ccc2ccccc12 |
| IUPAC Name | 1-aminonaphthalen-2-ol;hydrochloride |
| Molecular Weight (g/mol) | 195.64 |
| Formula Weight | 195.0451 |