Learn More
1-Amino-2-methylindoline, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Amino-2-methylindoline |
| CAS | 31529-46-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 H12 N2 |
| Quantity | 1 g |
| Synonym | 1H-Indol-1-amine, 2,3-dihydro-2-methyl-, Indoline, 1-amino-2-methyl- (6CI,8CI), 2,3-Dihydro-2-methyl-1H-indol-1-amine, 1-Amino-2-methylindoline, N-Amino-2-methylindoline, (2RS)-2-Methyl-2,3-dihydro-1H-indol-1-amine |
| InChI Formula | InChI=1S/C9H12N2/c1-7-6-8-4-2-3-5-9(8)11(7)10/h2-5,7H,6,10H2,1H3 |
| SMILES | CC1Cc2ccccc2N1N |
| IUPAC Name | 2-methyl-2,3-dihydroindol-1-amine |
| Molecular Weight (g/mol) | 148.2 |
| Show More |