missing translation for 'onlineSavingsMsg'
Learn More

1,6-Naphthyridine-2-carboxylic acid, 98%, Thermo Scientific™

Product Code. p-7077683 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15535256 250 mg 250mg
15525256 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535256 Supplier Thermo Scientific Acros Supplier No. 457532500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 197507-59-8
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.16
InChI Key OZZMWXQJCJUCEJ-UHFFFAOYSA-N
Synonym 1,6-naphthyridine-2-carboxylicacid, 1,6 naphthyridine-2-carboxylic acid, peakdale1_000097, 2-1,6 naphthyridinecarboxylic acid, 2-carboxy-1,6-naphthyridine, pyridino 4,3-b pyridine-2-carboxylic acid
PubChem CID 735908
IUPAC Name 1,6-naphthyridine-2-carboxylic acid
SMILES C1=CC(=NC2=C1C=NC=C2)C(=O)O

Specifications

Melting Point 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 174.16
Percent Purity 98%
Chemical Name or Material 1,6-Naphthyridine-2-carboxylic acid
compliance-icons
Product Identifier
  • 1,6-Naphthyridine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.