Learn More
1,5-Dibromoanthracene, TRC
Product Code. 30348613
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348613 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30348613 Supplier Toronto Research Chemicals Supplier No. TRCB42645310MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,5-Dibromoanthracene
CAS 3278-82-8
Molecular Formula C14H8Br2
Quantity 10 mg
InChI Formula InChI=1S/C14H8Br2/c15-13-5-1-3-9-7-12-10(8-11(9)13)4-2-6-14(12)16/h1-8H
SMILES Brc1cccc2cc3c(Br)cccc3cc12
IUPAC Name 1,5-dibromoanthracene
Molecular Weight (g/mol) 336.02
Formula Weight 333.8993