Learn More
1,5-Diaminonaphthalene, TRC
Product Code. 17973485
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973485 1 g 1g
17932885 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17973485 Supplier Toronto Research Chemicals Supplier No. TRCD4164151G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2243-62-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.2
Synonym 1,5-Naphthalenediamine, 1,5-DAN, 1,5-Diaminonaphthalene, 1,5-Naphthylenediamine, NSC 401110
IUPAC Name naphthalene-1,5-diamine
SMILES Nc1cccc2c(N)cccc12
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H10N2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6H,11-12H2
Formula Weight 158.0844
Chemical Name or Material 1,5-Diaminonaphthalene