missing translation for 'onlineSavingsMsg'
Learn More

1,5-Bis(diphenylphosphino)pentane, 97%, Thermo Scientific™

Product Code. p-3756927 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10155330 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10155330 Supplier Thermo Scientific Acros Supplier No. 320850010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27721-02-4
Molecular Formula C29H30P2
Molecular Weight (g/mol) 440.51
MDL Number MFCD00003052
InChI Key MZFPAWGWFDGCHP-UHFFFAOYSA-N
Synonym 1,5-bis diphenylphosphino pentane, dpppe, dppent, 5-diphenylphosphanylpentyl diphenyl phosphane, pentamethylenebis diphenylphosphine, 1,5-pentanediylbis diphenylphosphine, phosphine, 1,5-pentanediylbis diphenyl, 1-5-diphenylphosphino pentyl phenyl phosphino benzene
PubChem CID 2733414
IUPAC Name 5-diphenylphosphanylpentyl(diphenyl)phosphane
SMILES C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42.0°C to 47.0°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 440.51
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 1, 5-Bis(diphenylphosphino)pentane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.