Learn More
1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethan-1-one, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethan-1-one |
| CAS | 937591-32-7 |
| Molecular Formula | C16 H22 B N O3 |
| Quantity | 25 mg |
| Synonym | 1-[2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indol-1-yl]ethanone, 1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone |
| InChI Formula | InChI=1S/C16H22BNO3/c1-11(19)18-9-8-12-10-13(6-7-14(12)18)17-20-15(2,3)16(4,5)21-17/h6-7,10H,8-9H2,1-5H3 |
| SMILES | CC(=O)N1CCc2cc(ccc12)B3OC(C)(C)C(C)(C)O3 |
| IUPAC Name | 1-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindol-1-yl]ethanone |
| Molecular Weight (g/mol) | 287.162 |
| Formula Weight | 287.169 |