Learn More
1,4-Dimethyl-5-phenyl-1H-pyrazol-3-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Dimethyl-5-phenyl-1H-pyrazol-3-ol |
| CAS | 39513-08-1 |
| Molecular Formula | C11H12N2O |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C11H12N2O/c1-8-10(13(2)12-11(8)14)9-6-4-3-5-7-9/h3-7H,1-2H3,(H,12,14) |
| SMILES | Cc1c(O)nn(C)c1c2ccccc2 |
| Molecular Weight (g/mol) | 188.23 |
| Formula Weight | 188.095 |