Learn More
1,4-Dichlorobutane-2S-3S-diol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Dichlorobutane-2S-3S-diol |
| CAS | 139165-54-1 |
| Molecular Formula | C4 H8 Cl2 O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C4H8Cl2O2/c5-1-3(7)4(8)2-6/h3-4,7-8H,1-2H2/t3-,4-/m1/s1 |
| SMILES | O[C@H](CCl)[C@H](O)CCl |
| IUPAC Name | (2S,3S)-1,4-dichlorobutane-2,3-diol |
| Molecular Weight (g/mol) | 159.01 |
| Formula Weight | 157.9901 |