missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromo-2,5-difluorobenzene, 98%

Product Code. p-7021776
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462377 5 g 5g
11472377 25 g 25g
11482377 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462377 Supplier Thermo Scientific Alfa Aesar Supplier No. A14248.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 327-51-5 | C6H2Br2F2 | 271.887 g/mol

1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2,5-didodecyl-4,4-biphenylylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromo-2,5-difluorobenzene has been used in the preparation of 1,2,4,5-tetrakis(phosphino)-3,6-difluorobenzenes and poly(2,5-difluoro-2′,5′-didodecyl-4,4′-biphenylylene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Stable under recommended storage conditions. Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-51-5
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00000346
InChI Key GLVMLJCMUBZVTJ-UHFFFAOYSA-N
Synonym 1,4-dibromo-2,5-difluoro-benzene, benzene, 1,4-dibromo-2,5-difluoro, 2,5-difluoro-1,4-dibromo benzene, maybridge1_003946, pubchem3441, acmc-1cmi9, chemwish ic06783, ksc495q1l, emolecules 493137, 2,5-dibromo-1,4-difluorobenzene
PubChem CID 67596
IUPAC Name 1,4-dibromo-2,5-difluorobenzene
SMILES C1=C(C(=CC(=C1Br)F)Br)F

Specifications

Melting Point 60°C to 64°C
Boiling Point 96°C (20 mmHg)
Quantity 5 g
Beilstein 2250095
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 271.89
Percent Purity 98%
Chemical Name or Material 1,4-Dibromo-2,5-difluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.