Learn More
1-(4-Chlorophenyl)-2-(ethylamino)propan-1-one Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloroethcathinone hydrochloride |
| CAS | 22198-75-0 |
| Molecular Formula | C11 H14 Cl N O . Cl H |
| Quantity | 10 mg |
| Synonym | 1-Propanone, 1-(4-chlorophenyl)-2-(ethylamino)-, hydrochloride (1:1), 1-Propanone, 1-(4-chlorophenyl)-2-(ethylamino)-, hydrochloride (9CI), Propiophenone, 4'-chloro-2-(ethylamino)-, hydrochloride (7CI,8CI), 4'-Chloro-2-(ethylamino)propiophenone hydrochloride, 1-(4-Chlorophenyl)-2-(ethylamino)propan-1-one hydrochloride, 4'-Chloroethcathinone hydrochloride, 4-Chloroethcathinone hydrochloride, 4-CEC hydrochloride |
| InChI Formula | InChI=1S/C11H14ClNO.ClH/c1-3-13-8(2)11(14)9-4-6-10(12)7-5-9;/h4-8,13H,3H2,1-2H3;1H |
| SMILES | Cl.CCNC(C)C(=O)c1ccc(Cl)cc1 |
| IUPAC Name | 1-(4-chlorophenyl)-2-(ethylamino)propan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 248.15 |
| Formula Weight | 247.0531 |