Learn More
1-(4-Chlorophenyl)-2-(ethylamino)propan-1-one Hydrochloride, TRC
Product Code. 30365849
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365849 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30365849 Supplier Toronto Research Chemicals Supplier No. TRCC3740362.5MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Chloroethcathinone hydrochloride
CAS 22198-75-0
Molecular Formula C11 H14 Cl N O . Cl H
Quantity 2.5 mg
Synonym 1-Propanone, 1-(4-chlorophenyl)-2-(ethylamino)-, hydrochloride (1:1), 1-Propanone, 1-(4-chlorophenyl)-2-(ethylamino)-, hydrochloride (9CI), Propiophenone, 4'-chloro-2-(ethylamino)-, hydrochloride (7CI,8CI), 4'-Chloro-2-(ethylamino)propiophenone hydrochloride, 1-(4-Chlorophenyl)-2-(ethylamino)propan-1-one hydrochloride, 4'-Chloroethcathinone hydrochloride, 4-Chloroethcathinone hydrochloride, 4-CEC hydrochloride
InChI Formula InChI=1S/C11H14ClNO.ClH/c1-3-13-8(2)11(14)9-4-6-10(12)7-5-9;/h4-8,13H,3H2,1-2H3;1H
SMILES Cl.CCNC(C)C(=O)c1ccc(Cl)cc1
IUPAC Name 1-(4-chlorophenyl)-2-(ethylamino)propan-1-one;hydrochloride
Molecular Weight (g/mol) 248.15
Formula Weight 247.0531