Learn More
1,4-Butane-d8-diamine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Diaminobutane-D8 Dihydrochloride |
| CAS | 284665-22-1 |
| Molecular Formula | C4 D8 H4 N2 . 2 Cl H |
| Quantity | 25 mg |
| Synonym | 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine, dihydrochloride (9CI), 1,4-Butane-d8-diamine dihydrochloride, 1,4-Diaminobutane D8 dihydrochloride, 1,4-Diaminobutane-D8 dihydrochloride, 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine dihydrochloride, 1,4-Butane-d8-diamine d |
| InChI Formula | InChI=1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H/i1D2,2D2,3D2,4D2;; |
| SMILES | Cl.Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C([2H])([2H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine;dihydrochloride |
| Molecular Weight (g/mol) | 169.123 |
| Formula Weight | 168.104 |