Learn More
1-(4-Bromophenyl)piperidin-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Bromophenyl)piperidin-2-one |
| CAS | 27471-43-8 |
| Molecular Formula | C11H12BrNO |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C11H12BrNO/c12-9-4-6-10(7-5-9)13-8-2-1-3-11(13)14/h4-7H,1-3,8H2 |
| SMILES | Brc1ccc(cc1)N2CCCCC2=O |
| IUPAC Name | 1-(4-bromophenyl)piperidin-2-one |
| Molecular Weight (g/mol) | 254.12 |
| Formula Weight | 253.0102 |