Learn More
1,4-Bis(maleimido)butane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Bis-maleimidobutane |
| CAS | 28537-70-4 |
| Molecular Formula | C12 H12 N2 O4 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H12N2O4/c15-9-3-4-10(16)13(9)7-1-2-8-14-11(17)5-6-12(14)18/h3-6H,1-2,7-8H2 |
| SMILES | O=C1C=CC(=O)N1CCCCN2C(=O)C=CC2=O |
| IUPAC Name | 1-[4-(2,5-dioxopyrrol-1-yl)butyl]pyrrole-2,5-dione |
| Molecular Weight (g/mol) | 248.23 |
| Formula Weight | 248.0797 |