Learn More
1,4-bis(7-Chloroquinolin-4-yl)piperazine, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-bis(7-Chloroquinolin-4-yl)piperazine |
| CAS | 31502-87-1 |
| Molecular Formula | C22H18Cl2N4 |
| Quantity | 250 mg |
| Synonym | 4,4'-(1,4-Piperazinediyl)bis[7-chloroquinoline] |
| InChI Formula | InChI=1S/C22H18Cl2N4/c23-15-1-3-17-19(13-15)25-7-5-21(17)27-9-11-28(12-10-27)22-6-8-26-20-14-16(24)2-4-18(20)22/h1-8,13-14H,9-12H2 |
| SMILES | C1=CC2=C(C=C1Cl)N=CC=C2N3CCN(CC3)C4=CC=NC5=C4C=CC(=C5)Cl |
| IUPAC Name | 7-chloro-4-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]quinoline |
| Molecular Weight (g/mol) | 409.31 |
| Formula Weight | 408.0909 |