Learn More
1,4-Bis[3,4-dihydro-2(1H)-quinolinon-7-oxy]butane-d8, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Bis[3,4-dihydro-2(1H)-quinolinon-7-oxy]butane-d8 |
| CAS | 1346599-01-6 |
| Molecular Formula | C22H16D8N2O4 |
| Quantity | 100 mg |
| Synonym | 7,7'-[1,4-(Butane-d8)diylbis(oxy)]bis[3,4-dihydro-2(1H)-quinolinone |
| InChI Formula | InChI=1S/C22H24N2O4/c25-21-9-5-15-3-7-17(13-19(15)23-21)27-11-1-2-12-28-18-8-4-16-6-10-22(26)24-20(16)14-18/h3-4,7-8,13-14H,1-2,5-6,9-12H2,(H,23,25)(H,24,26)/i1D2,2D2,11D2,12D2 |
| SMILES | [2H]C([2H])(Oc1ccc2CCC(=O)Nc2c1)C([2H])([2H])C([2H])([2H])C([2H])([2H])Oc3ccc4CCC(=O)Nc4c3 |
| IUPAC Name | 7-[1,1,2,2,3,3,4,4-octadeuterio-4-[(2-oxo-3,4-dihydro-1H-quinolin-7-yl)oxy]butoxy]-3,4-dihydro-1H-quinolin-2-one |
| Molecular Weight (g/mol) | 388.49 |
| Formula Weight | 388.2238 |